Learn More
Medchemexpress LLC FAPy-adenine | 5122-36-1 | 99.9% | 153.14 | 50 MG
SDP
Catalog No. 5000436359
Encompass_Preferred
FAPy-adenine is an oxidized DNA base. Increased levels of FAPy-adenine have been observed in the brains of individuals with Alzheimer's disease.
  • Oxidized nucleosides are biochemical markers for various conditions, including tumors, aging, and neurodegenerative diseases.
  • Intended for research use only and not for patient use.
Catalog No. 50-004-36359 Supplier Medchemexpress LLC Supplier No. HY11330350MG
FAPy-adenine is an oxidized DNA base. Increased levels of FAPy-adenine have been observed in the brains of individuals with Alzheimer's disease.
  • Oxidized nucleosides are biochemical markers for various conditions, including tumors, aging, and neurodegenerative diseases.
  • Intended for research use only and not for patient use.