Description
- Shields keyfobs, Apple Watch, WiFi cards, thumb drives, and other small electronics while blocking WiFi, Bluetooth, cellular, GPS, RFID, and radio signals with an average attenuation of 60 to 80 dB.
- Provides EMI and RFI shielding to support vehicle security and data protection for small electronic devices.
- Features a keyring loop and belt loop for convenient carrying.
- Constructed with two layers of High-shielding fabric.
- Supplied in packs of 2 units.
Specifications
Specifications
| Dimensions (L x W) | 5 x 3.25 in. |
| Product Type | Evidence Bag |
| Color | Black |