Learn More
Medchemexpress LLC Fargesin | 31008-19-2 | 370.40 | 5 MG
SDP
Catalog No. 5000416804
Encompass_Preferred
Fargesin is a bioactive neolignan isolated from magnolia plants, known for its antihypertensive and anti-inflammatory effects. It is intended for research use only and is not sold to patients.
  • Bioactive neolignan
  • Isolated from magnolia plants
  • Exhibits antihypertensive and anti-inflammatory effects
  • Suppresses the PKC pathway in THP-1 cells (including JNK, AP-1, and NF-κB)
  • Acts as a potential β₁ adrenergic receptor antagonist
  • Protects hearts against ischemia/reperfusion injury in rats
  • Attenuates oxidative stress and apoptosis
  • Promotes NO release in 2K1C hypertensive rats
Catalog No. 50-004-16804 Supplier Medchemexpress LLC Supplier No. HYN07195MG
Fargesin is a bioactive neolignan isolated from magnolia plants, known for its antihypertensive and anti-inflammatory effects. It is intended for research use only and is not sold to patients.
  • Bioactive neolignan
  • Isolated from magnolia plants
  • Exhibits antihypertensive and anti-inflammatory effects
  • Suppresses the PKC pathway in THP-1 cells (including JNK, AP-1, and NF-κB)
  • Acts as a potential β₁ adrenergic receptor antagonist
  • Protects hearts against ischemia/reperfusion injury in rats
  • Attenuates oxidative stress and apoptosis
  • Promotes NO release in 2K1C hypertensive rats