Learn More
Medchemexpress LLC Fargesin | 31008-19-2 | 99.5% | 370.40 | C21H22O6 | 10 MG
SDP
Catalog No. 5000295627
Encompass_Preferred
Fargesin is a bioactive neolignan isolated from Magnolia species, supplied as a research-grade analytical standard. It exhibits reported antihypertensive and anti-inflammatory activity and is provided as a solid powder with high purity for laboratory use. Refer to the SDS for handling and storage.
  • Bioactive neolignan with reported antihypertensive and anti-inflammatory effects.
  • High purity (~99.5%) analytical standard suitable for research applications.
  • Solid powder supplied in a 10 mg vial.
  • Molecular formula C21H22O6; molecular weight 370.40 g·mol-1.
  • Store under cold conditions and follow the SDS for handling.
Catalog No. 50-002-95627 Supplier Medchemexpress LLC Supplier No. HYN071910MG
Fargesin is a bioactive neolignan isolated from Magnolia species, supplied as a research-grade analytical standard. It exhibits reported antihypertensive and anti-inflammatory activity and is provided as a solid powder with high purity for laboratory use. Refer to the SDS for handling and storage.
  • Bioactive neolignan with reported antihypertensive and anti-inflammatory effects.
  • High purity (~99.5%) analytical standard suitable for research applications.
  • Solid powder supplied in a 10 mg vial.
  • Molecular formula C21H22O6; molecular weight 370.40 g·mol-1.
  • Store under cold conditions and follow the SDS for handling.