Learn More
Grainger Solo Handheld Sprayer, 3 Gal., HDPE Tank, Viton Seals, 1 ea.
Catalog No. 502786318
Encompass_Preferred
This 3 gallon handheld sprayer is designed for medium-to-large spraying tasks at home, on the farm, or in professional landscaping and agricultural environments. Built with a durable, high-density polyethylene (HDPE) tank, the unit is corrosion and ultraviolet resistant. It features chemical-resistant Viton® seals in key locations to withstand harsh substances, making it suitable for fertilizers, herbicides, and pesticides.
  • Includes a 4-nozzle assortment: plastic adjustable, fan spray, hollow cone, and jet stream nozzles; compatible with TeeJet® nozzle tips
  • Features a 28 in. unbreakable wand and a commercial-quality shut-off valve with a lock-on/lock-off feature
  • Equipped with a funnel top for easy filling and a large secondary opening for convenient cleaning
  • Includes a built-in storage compartment for extra nozzles
  • Rugged piston pump allows for quick pressurization
Catalog No. 50-278-6318 Supplier Grainger Supplier No. 190T24
This 3 gallon handheld sprayer is designed for medium-to-large spraying tasks at home, on the farm, or in professional landscaping and agricultural environments. Built with a durable, high-density polyethylene (HDPE) tank, the unit is corrosion and ultraviolet resistant. It features chemical-resistant Viton® seals in key locations to withstand harsh substances, making it suitable for fertilizers, herbicides, and pesticides.
  • Includes a 4-nozzle assortment: plastic adjustable, fan spray, hollow cone, and jet stream nozzles; compatible with TeeJet® nozzle tips
  • Features a 28 in. unbreakable wand and a commercial-quality shut-off valve with a lock-on/lock-off feature
  • Equipped with a funnel top for easy filling and a large secondary opening for convenient cleaning
  • Includes a built-in storage compartment for extra nozzles
  • Rugged piston pump allows for quick pressurization