Learn More
Medchemexpress LLC Farnesene (mixture of isomers) | 125037-13-0 | 98.0% | 204.35 | 1 ML
SDP
Catalog No. 5000360744
Encompass_Preferred
Farnesene (mixture of isomers) is a farnesene with a mixture of isomers. It is a herbivore-induced plant volatile (HIPV) and plays a significant role in insect resistance across various plant species. It is for research use only.
  • Exists as a mixture of isomers
  • Herbivore-induced plant volatile (HIPV)
  • Important effect on insect resistance in many plant species
Catalog No. 50-003-60744 Supplier Medchemexpress LLC Supplier No. HY14620A10MM/1ML
Farnesene (mixture of isomers) is a farnesene with a mixture of isomers. It is a herbivore-induced plant volatile (HIPV) and plays a significant role in insect resistance across various plant species. It is for research use only.
  • Exists as a mixture of isomers
  • Herbivore-induced plant volatile (HIPV)
  • Important effect on insect resistance in many plant species