Learn More
Sigma Aldrich Fine Chemicals Biosciences farnesene mixture of isome100G
Catalog No. 502901744
Encompass
farnesene (mixture of isomers) may contain sequiterpenes trans--farnesene cis -farnesene trans -farnesene and bisabolene. farnesene is reported to be an oviposition stimulant for Laspeyresia pomonella while trans--farnesene as a feeding stimulant for Lutzomyia longipalpis.
Catalog No. 50-290-1744 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W383902100GK
farnesene (mixture of isomers) may contain sequiterpenes trans--farnesene cis -farnesene trans -farnesene and bisabolene. farnesene is reported to be an oviposition stimulant for Laspeyresia pomonella while trans--farnesene as a feeding stimulant for Lutzomyia longipalpis.