Learn More
Electron Microscopy Sciences Farnesol 10 ML
SDP
Catalog No. 50980469
Encompass_Preferred
(3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol)
CAS #4602-84-0 C₁₅H₂₆O F.W. 222.57
Mixture of Stereoisomers: Approx. 90% Synthetic
Catalog No. 50-980-469 Supplier Electron Microscopy Sciences Supplier No. 15340
Add to Cart
Edge
Add to Cart
(3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol)
CAS #4602-84-0 C₁₅H₂₆O F.W. 222.57
Mixture of Stereoisomers: Approx. 90% Synthetic