Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Farnesol, 97.7%, MP Biomedicals™

Catalog No. ICN10168005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10168005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10168005 Supplier MP Biomedicals, Inc Supplier No. 0210168005

Chemical Identifiers

CAS 4602-84-0
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
Synonym e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
PubChem CID 1549109
ChEBI CHEBI:35966
IUPAC Name (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO

Specifications

Color Colorless
Density 0.886g/mL
Assay Percent Range 97.7%
Quantity 5 mL
Formula Weight 222.36
Physical Form Liquid
Chemical Name or Material Farnesol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.