Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Farnesol, Mixture of Isomers 96%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
4602-84-0
C15H26O
222.37
MFCD00002918
CRDAMVZIKSXKFV-GNESMGCMSA-N
e,z-farnesol,2-trans,6-cis-farnesol,2e,6z-farnesol,trans,cis-farnesol,farnesol,2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol,farnesyl alcohol,e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol,2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
1549109
CHEBI:35966
(2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
CC(=CCCC(=CCCC(=CCO)C)C)C
4602-84-0
C15H26O
222.37
MFCD00002918
CRDAMVZIKSXKFV-GNESMGCMSA-N
e,z-farnesol,2-trans,6-cis-farnesol,2e,6z-farnesol,trans,cis-farnesol,farnesol,2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol,farnesyl alcohol,e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol,2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
1549109
CHEBI:35966
(2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
CC(=CCCC(=CCCC(=CCO)C)C)C
Boiling Point 149°C (4.0mmHg)
Flash Point 96°C
Specific Gravity 0.8865
Refractive Index 1.4870 to 1.492
Density 0.8865g/mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 25mL
Packaging Glass bottle
Assay Percent Range 96%
Beilstein 01, III, 2040
Merck Index 15, 3975
Solubility Information Solubility in water: immiscible
Formula Weight 222.37
Physical Form Liquid
Percent Purity ≥95% (sum of geometric isomers)
Chemical Name or Material Farnesol, 96%
Show More Show Less

EINECSNumber : 225-004-1

RTECSNumber : JR4979000

TSCA : TSCA