Learn More
Medchemexpress LLC Farnesol (standard) | 106-28-5 | MFCD00002918 | 99.2% | 222.37 | C15H26O | 5g
SDP
Catalog No. 5000267957
Encompass_Preferred
(E E)-Farnesol (trans trans-Farnesol) is a quorum-sensing molecule of Candida species (E E)-Farnesol can inhibit the growth metabolism and biofilm formation of various Candida species and affect their morphology and invasiveness[1]
Catalog No. 50-002-67957 Supplier Medchemexpress LLC Supplier No. HYY02485G
(E E)-Farnesol (trans trans-Farnesol) is a quorum-sensing molecule of Candida species (E E)-Farnesol can inhibit the growth metabolism and biofilm formation of various Candida species and affect their morphology and invasiveness[1]