Learn More
Medchemexpress LLC Farnesyl pyrophosphate ammonium | 116057-57-9 | 95.39% | 433.42 | 1 MG
SDP
Catalog No. 5000439620
Encompass_Preferred
Farnesyl pyrophosphate ammonium is a metabolic intermediate in the mevalonate (MVA) pathway. It acts as a TRP channel (TRPM2) agonist, triggering Ca2+ influx and cell death. It is a crucial branch substrate for various biological syntheses and is utilized in research concerning a range of diseases.
  • Acts as a TRP channel (TRPM2) agonist
  • Triggers Ca2+ influx and cell death
  • Key branch substrate for cholesterol synthesis
  • Key branch substrate for ubiquinone synthesis
  • Key branch substrate for protein farnesylation
  • Key branch substrate for geranylgeranyl pyrophosphate (GGPP) synthesis
  • Used in research on cerebral ischemia
  • Used in research on neurodegenerative diseases
  • Used in research on pancreatic cancer
  • Used in research on inflammation
  • Used in research on autoimmune diseases
Catalog No. 50-004-39620 Supplier Medchemexpress LLC Supplier No. HY113037C1MG
Farnesyl pyrophosphate ammonium is a metabolic intermediate in the mevalonate (MVA) pathway. It acts as a TRP channel (TRPM2) agonist, triggering Ca2+ influx and cell death. It is a crucial branch substrate for various biological syntheses and is utilized in research concerning a range of diseases.
  • Acts as a TRP channel (TRPM2) agonist
  • Triggers Ca2+ influx and cell death
  • Key branch substrate for cholesterol synthesis
  • Key branch substrate for ubiquinone synthesis
  • Key branch substrate for protein farnesylation
  • Key branch substrate for geranylgeranyl pyrophosphate (GGPP) synthesis
  • Used in research on cerebral ischemia
  • Used in research on neurodegenerative diseases
  • Used in research on pancreatic cancer
  • Used in research on inflammation
  • Used in research on autoimmune diseases