Learn More
Medchemexpress LLC Farnesyl pyrophosphate-d2 triammonium | 00-00-0 | MFCD00010901 | 95.8% | 435.43 g/mol | C15H35D2N3O7P2 | 5 MG
SDP
Catalog No. 5000262444
Encompass_Preferred
Farnesyl pyrophosphate-d2 triammonium is a deuterium-labeled form of farnesyl pyrophosphate supplied as the triammonium salt. It functions as a metabolic intermediate in the mevalonate pathway, acts as a TRPM2 agonist in cellular assays, and is used as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses in biochemical research related to cholesterol, ubiquinone, and protein prenylation pathways.
  • Deuterium-labeled tracer for quantitative analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Acts as a TRPM2 agonist in cellular assays.
  • High purity for reliable analytical results.
  • Solid, white to off-white form for stable storage and handling.
Catalog No. 50-002-62444 Supplier Medchemexpress LLC Supplier No. HY113037CS5MG
Farnesyl pyrophosphate-d2 triammonium is a deuterium-labeled form of farnesyl pyrophosphate supplied as the triammonium salt. It functions as a metabolic intermediate in the mevalonate pathway, acts as a TRPM2 agonist in cellular assays, and is used as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses in biochemical research related to cholesterol, ubiquinone, and protein prenylation pathways.
  • Deuterium-labeled tracer for quantitative analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Acts as a TRPM2 agonist in cellular assays.
  • High purity for reliable analytical results.
  • Solid, white to off-white form for stable storage and handling.