Learn More
Echelon Biosciences Research Labs FARNESYL S-DIPHOSPHATE 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs IS15010MG
Encompass
Farnesyl thiolodiphosphate (FSPP) is an analog of FPP in which the non-bridging oxygen has been replaced by sulfur resulting in a less-reactive compound. This item is non returnable
Catalog No. 50-720-415