Learn More
Cayman Chemical Farnesyl Thiosalicylic Acid, Purity: ≥96%, CAS: 162520-00-5, 5 mg
SDP
Catalog No. 502890694
Encompass_Preferred
An inhibitor of Ras-mediated signaling that functions by dislodging Ras from the cell membrane thereby rendering it susceptible to proteolytic degradation; inhibits the growth of human Ha-ras-transformed Rat1 fibroblasts with an IC50 value of 7.5 µM
Catalog No. 50-289-0694 Supplier Cayman Chemical Supplier No. 100105015 MG
An inhibitor of Ras-mediated signaling that functions by dislodging Ras from the cell membrane thereby rendering it susceptible to proteolytic degradation; inhibits the growth of human Ha-ras-transformed Rat1 fibroblasts with an IC50 value of 7.5 µM