Learn More
Enzo Life Sciences Farnesyl thiotriazole (10mg). CAS: 156604-45-4
SDP
Supplier:  Enzo Life Sciences BMLPE1900010
Encompass_Preferred
A structurally unique activator of PKC in whole cells or in purified enzyme preparations (Kd=2.5 µM) which is equipotent with dioleoylglycerol. Since it does not possess hydrolyzable esters of fatty acids, it represents a stable, persistent PKC activator. Alternative name: FTT. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-1409