Learn More
Cayman Chemical Farnesene (mixed isomers), 25 g
SDP
Catalog No. 502934622
Encompass_Preferred
A mixture of farnesene isomers; may contain trans-β-farnesene, cis-α-farnesene, trans-α-farnesene, and bisabolene, among others
Catalog No. 50-293-4622 Supplier Cayman Chemical Supplier No. 2689125 G
A mixture of farnesene isomers; may contain trans-β-farnesene, cis-α-farnesene, trans-α-farnesene, and bisabolene, among others