Learn More
Cayman Chemical Farnesene (mixed isomers), 50 g
SDP
Catalog No. 502935006
Encompass_Preferred
A mixture of farnesene isomers; may contain trans-β-farnesene, cis-α-farnesene, trans-α-farnesene, and bisabolene, among others
Catalog No. 50-293-5006 Supplier Cayman Chemical Supplier No. 2689150 G
A mixture of farnesene isomers; may contain trans-β-farnesene, cis-α-farnesene, trans-α-farnesene, and bisabolene, among others