Learn More
Cayman Chemical Farnsyl ThIosalcylc ACD A 10mg
SDP
Catalog No. 502953447
Encompass_Preferred
An inhibitor of Ras-mediated signaling that inhibits the growth of PANC-1 and U87 tumor cells with IC50 values of 20 and 10 µM, respectively
Catalog No. 50-295-3447 Supplier Cayman Chemical Supplier No. 1317510 MG
An inhibitor of Ras-mediated signaling that inhibits the growth of PANC-1 and U87 tumor cells with IC50 values of 20 and 10 µM, respectively