Learn More
Cayman Chemical Farnsyl ThIosalcylc ACD A 50mg
SDP
Catalog No. 502951750
Encompass_Preferred
An inhibitor of Ras-mediated signaling that inhibits the growth of PANC-1 and U87 tumor cells with IC50 values of 20 and 10 µM, respectively
Catalog No. 50-295-1750 Supplier Cayman Chemical Supplier No. 1317550 MG
An inhibitor of Ras-mediated signaling that inhibits the growth of PANC-1 and U87 tumor cells with IC50 values of 20 and 10 µM, respectively