Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Faropenem Sodium, G-Biosciences
SCI_ED

Catalog No. S139818
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139818 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139818 Supplier G-Biosciences Supplier No. RC1020
Add to Cart
Add to Cart

Faropenem sodium is an ultra broad-spectrum Beta-lactam of the penem antibiotic class.

It has been shown to be Beta-lactamase resistant and is especially effective against anaerobic bacteria responsible for oral infections, but targets a variety of gram-positive and gram-negative bacteria. Faropenem sodium is freely soluble in water.

Chemical Identifiers

Molecular Weight (g/mol) 307.30 g/mol

Specifications

Boiling Point 570.2°C
Quantity 50 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.