Learn More
Cayman Chemical FaropenmsodIM salt hydrat 50mg
SDP
Catalog No. 502950678
Encompass_Preferred
A penem antibiotic; active against a panel of 12 Gram-positive (MIC50s = 0.008-1 mg/L) and 15 Gram-negative (MIC50s = 0.06-16 mg/L) bacteria; reduces the number of lung CFUs in a mouse model of H. influenzae-induced respiratory tract infection
Catalog No. 50-295-0678 Supplier Cayman Chemical Supplier No. 3450750 MG
A penem antibiotic; active against a panel of 12 Gram-positive (MIC50s = 0.008-1 mg/L) and 15 Gram-negative (MIC50s = 0.06-16 mg/L) bacteria; reduces the number of lung CFUs in a mouse model of H. influenzae-induced respiratory tract infection