Learn More
Aviva Systems Biology Corp FARS2 Antibody - N-terminal region (ARP40669_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40669_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FARS2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FARS2. Alternate names: FARS1, HSPC320, PheRS, dJ520B18.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57581