Learn More
Medchemexpress LLC Farudodstat (ASLAN003) | 1035688-66-4 | 99.7% | 100 MG
SDP
Catalog No. 5000338889
Encompass_Preferred
Farudodstat (ASLAN003) is an orally active and potent Dihydroorotate Dehydrogenase (DHODH) inhibitor with an IC50 of 35 nM for the human DHODH enzyme. Its mechanism of action involves inhibiting protein synthesis by activating AP-1 transcription factors. This compound induces apoptosis and significantly prolongs survival in acute myeloid leukemia (AML) xenograft mice.
  • Exhibits antiproliferative activity against various human cell lines.
  • Inhibits leukemic cell proliferation in vitro.
  • Increases cleaved caspase 8.
  • Decreases viability and induces differentiation in primary acute myeloid leukemia blasts and myelodysplastic syndrome samples.
  • Inhibits protein synthesis by reducing O-propargyl-puromycin (OPP) incorporation and downregulating EIF4B and RPL6 proteins.
  • Substantially reduces the number and size of disseminated tumors in vivo.
  • Available as a white to off-white solid.
Catalog No. 50-003-38889 Supplier Medchemexpress LLC Supplier No. HY129239100MG
Farudodstat (ASLAN003) is an orally active and potent Dihydroorotate Dehydrogenase (DHODH) inhibitor with an IC50 of 35 nM for the human DHODH enzyme. Its mechanism of action involves inhibiting protein synthesis by activating AP-1 transcription factors. This compound induces apoptosis and significantly prolongs survival in acute myeloid leukemia (AML) xenograft mice.
  • Exhibits antiproliferative activity against various human cell lines.
  • Inhibits leukemic cell proliferation in vitro.
  • Increases cleaved caspase 8.
  • Decreases viability and induces differentiation in primary acute myeloid leukemia blasts and myelodysplastic syndrome samples.
  • Inhibits protein synthesis by reducing O-propargyl-puromycin (OPP) incorporation and downregulating EIF4B and RPL6 proteins.
  • Substantially reduces the number and size of disseminated tumors in vivo.
  • Available as a white to off-white solid.