Learn More
Aviva Systems Biology Corp FAS Antibody - middle region (ARP30627_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30627_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAS. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FAS. Alternate names: ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-53946