Learn More
Enzo Life Sciences Fas (human) monoclonal antibody (blocking) (SM1/23) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805010C100
Encompass_Preferred
Clone: SM1/23. Isotype: IgG2b. Immunogen: Human Fas (CD95; APO-1). Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2-7.4.
Catalog No. 50-201-0029