Learn More
AdipoGen anti-Fas (human), mAb (APO-1-1) (preservative free) (500 µg)
SDP
Supplier:  AdipoGen AG20B0079PFC500
Encompass_Preferred
Monoclonal antibody. Recognizes human Fas. Applications: FACS, FUNC, IHC. Clone: APO-1-1. Isotype: Mouse IgG1. Liquid. In PBS. Purity: >95% (SDS-PAGE). Fas (CD95) is a member of the death receptor (DR) family, a subfamily of the tumor necrosis factor receptor superfamily. The formation of the Fas death-inducing signaling complex (DISC) is the initial step of Fas signaling. Activation of procaspase-8 at the DISC leads to the induction of DR-mediated apoptosis. Stimulation of Fas has also been reported to trigger non-apoptotic pathways. It has been shown that membrane-bound FasL is essential for the cytotoxic activity, whereas soluble FasL appears to promote autoimmunity and tumorigenesis via induction of non-apoptotic pathways, in particular NF-kappaB.
Catalog No. 50-205-2994