Learn More
Enzo Life Sciences Fas (human) monoclonal antibody (2R2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805038C100
Encompass_Preferred
Clone: 2R2. Isotype: IgG3. Immunogen: Mouse L cells stably overexpressing human Fas (CD95; APO-1). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 1% BSA. Contains no preservatives.
Catalog No. 50-201-0039