Learn More
Enzo Life Sciences Fas (human) monoclonal antibody (5H5) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7130100
Encompass_Preferred
Clone: 5H5. Isotype: IgG1κ. Immunogen: Human recombinant Fas (CD95; APO-1). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Applications: ELISA, Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS. Contains no preservatives.
Catalog No. 50-201-3251