Learn More
Enzo Life Sciences Fas (human) monoclonal antibody (APO-1-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805022C100
Encompass_Preferred
Clone: APO-1-1. Isotype: IgG1. Immunogen: Human Fas (CD95; APO-1). Species reactivity: Human. Applications: Flow Cytometry, IHC (FS), FUNC. Formulation: Liquid. In PBS, pH 7.2-7.4.
Catalog No. 50-201-0033