Learn More
Enzo Life Sciences Fas polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP221D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse Fas. UniProt ID: P25446. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (1:50) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7156