Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6624MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: FSCN1/417. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HeLa, MCF-7, PC-3 or BEWO cells. Hodgkin.’s lymphoma, Ovarian or Testicular Carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Full length recombinant human FSCN1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55kDa.Chromosome: 7p22.1. Gene Symbol: FSCN1. Gene ID: 6624. Ensemble: ENSG00000075618. Uniprot: NP_003079.
Catalog No. 50-188-5176