Learn More
Drg International Inc DRG Fasciola IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA4503R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Fasciola in serum and plasma (EDTA-, or heparin- plasma). This product is for Research Use Only. The DRG Fasciola IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Fasciola antigen from adult liver fluke. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7611