Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fasentin, Tocris Bioscience™

Catalog No. 610010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 61-001-0 Supplier R&D Systems Supplier No. 6100/10
This item is not returnable. View return policy

GLUT1/GLUT4 inhibitor; also Fas-sensitizer

GLUT1 and GLUT4 inhibitor (IC50 = 68 μM). Preferentially inhibits GLUT4 over GLUT1. Sensitizes Fas receptor in a range of tumor cell lines (IC50 = 20 μM) by modulating the extrinsic apoptotic pathway downstream of TNF receptors but upstream of effector caspases.

Chemical Identifiers

CAS 392721-37-8
Molecular Formula C11H9ClF3NO2
Molecular Weight (g/mol) 279.64
MDL Number MFCD01001285
InChI Key GNYIJZMBLZXJEJ-UHFFFAOYSA-N
Synonym Fasentin
PubChem CID 879520
IUPAC Name N-[4-chloro-3-(trifluoromethyl)phenyl]-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC(=C(Cl)C=C1)C(F)(F)F

Specifications

Linear Formula C11H9ClF3NO2
Quantity 10 mg
Solubility Information Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 279.64
Percent Purity >98%
Physical Form Solid
Chemical Name or Material N-(4-Chloro-3-(trifluoromethyl)phenyl)-3-oxobutanamide
Recommended Storage Store at −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.