Learn More
ACROBiosystems Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein, Fc Tag;HEK293;100UG;FAS,FAS,ALPS1A,APO-1,APO1,APT1,APT-1,CD95,CD-95,FAS1,FAS-1,FASTM,TNFRSF6,TNFRSF-6,FasR ,Fas-R
Supplier:  ACROBiosystems FASH5252100UG
Encompass_Preferred
Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein, Fc Tag;100UG;Human Fas, Fc Tag (FAS-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Asn 173 (Accession # AAH12479.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.8 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Fas Ligand, His Tag (Cat. No. FAL-H5241) at 5μg/mL (100 μL/well) can bind Human Fas, Fc Tag (Cat. No. FAS-H5252) with a linear range of 1.56-12.5 ng/mL (QC tested).
Catalog No. 50-165-7576