Learn More
Medchemexpress LLC Fasiglifam (TAK-875) | 1000413-72-8 | MFCD18251445 | 99.0% | 524.63 | C29H32O7S | 200 MG
SDP
Catalog No. 5000244183
Encompass_Preferred
Fasiglifam (TAK-875) is a potent, selective, orally bioavailable GPR40 (FFAR1) agonist used in research to study glucose-dependent insulin secretion and GPR40 pharmacology. The compound is supplied as a solid research reagent with documented potency and chemical characterization (formula C29H32O7S; MW 524.63 g/mol).
  • Potent, selective GPR40 agonist with reported EC50 of 72 nM.
  • Orally bioavailable pharmacology profile suitable for in vitro and in vivo studies.
  • High purity (~99%) suitable for laboratory research.
  • Available in multiple sizes including milligram and gram quantities.
  • Chemical formula C29H32O7S and molecular weight 524.63 g/mol.
Catalog No. 50-002-44183 Supplier Medchemexpress LLC Supplier No. HY10480200MG
Fasiglifam (TAK-875) is a potent, selective, orally bioavailable GPR40 (FFAR1) agonist used in research to study glucose-dependent insulin secretion and GPR40 pharmacology. The compound is supplied as a solid research reagent with documented potency and chemical characterization (formula C29H32O7S; MW 524.63 g/mol).
  • Potent, selective GPR40 agonist with reported EC50 of 72 nM.
  • Orally bioavailable pharmacology profile suitable for in vitro and in vivo studies.
  • High purity (~99%) suitable for laboratory research.
  • Available in multiple sizes including milligram and gram quantities.
  • Chemical formula C29H32O7S and molecular weight 524.63 g/mol.