Learn More
AdipoGen FasL (human) (multimeric) (rec.) MultiPack,Protein
SDP
Supplier:  AdipoGen AG40B01303010
Encompass_Preferred
FasL (human) (multimeric) (rec.) MultiPack, AdipoGen Life Sciences. Protein. Human FasL (aa 139-281) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG¿-tag. Source: HEK 293 cells. Endotoxin content: <0.02EU/¿g purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse Fas. Purity: >95% (SDS-PAGE). MultimericFasL¿ is a high activity construct in which two trimeric FasL are artificially linked via the collagen domain of ACRP30. This construct very effectively mimics the natural membrane-assisted aggregation of FasL in vivo. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. It is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Catalog No. 50-594-439