Learn More
Enzo Life Sciences FasL (mouse) monoclonal antibody (H11) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804010C100
Encompass_Preferred
Clone: H11. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 196-220 of mouse FasL (CD95L; APO-1L; CD178). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Mouse. Applications: Flow Cytometry, ICC, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9744