Learn More
AdipoGen FasL Soluble hrec
SDP
Catalog No. 502562942
Encompass_Preferred
Protein. The extracellular domain of human FasL aa 103-281 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.05EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. Is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Catalog No. 50-256-2942 Supplier AdipoGen Supplier No. AG40B00013010
Protein. The extracellular domain of human FasL aa 103-281 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.05EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. Is involved in cytotoxic T cell mediated apoptosis and in T cell development.