Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fatty acid synthase (human), recombinant, His-tagged | 9045-77-6 | 50UG
SDP

Catalog No. 5000176384
Encompass_Preferred

Recombinant human fatty acid synthase (FASN) expressed in Sf9 insect cells and supplied as a His-tagged, 0.22 μm filtered protein solution suitable for biochemical and structural studies.

  • His tag for affinity purification and detection.
  • Molecular weight 47.2 kDa.
  • Formulated in 50 mM HEPES, 200 mM NaCl, 10% glycerol, 1 mM DTT, pH 7.5.
  • Endotoxin <1 EU/μg (LAL method).
  • Stored at -80°C; stable at -20°C for 3 months after opening.
  • Avoid repeated freeze-thaw; aliquot for extended storage.

Catalog No. 50-001-76384 Supplier Medchemexpress LLC Supplier No. HYP70188250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human fatty acid synthase (FASN) expressed in Sf9 insect cells and supplied as a His-tagged, 0.22 μm filtered protein solution suitable for biochemical and structural studies.

  • His tag for affinity purification and detection.
  • Molecular weight 47.2 kDa.
  • Formulated in 50 mM HEPES, 200 mM NaCl, 10% glycerol, 1 mM DTT, pH 7.5.
  • Endotoxin <1 EU/μg (LAL method).
  • Stored at -80°C; stable at -20°C for 3 months after opening.
  • Avoid repeated freeze-thaw; aliquot for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.