Learn More
Medchemexpress LLC HY-111777 10mg Medchemexpress, FASN-IN-1 CAS:1808260-84-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11177710MG
Encompass_Preferred
Medchemexpress, HY-111777 10mg FASN-IN-1 CAS:1808260-84-5 FASN-IN-1 is a fatty acid synthase (FASN) inhibitor extracted from patent WO2015134790A1, compound 56. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8959