Learn More
Medchemexpress LLC HY-111777 5mg Medchemexpress, FASN-IN-1 CAS:1808260-84-5 Purity:>98%
SDP
Catalog No. 501932391
Encompass_Preferred
Medchemexpress, HY-111777 5mg FASN-IN-1 CAS:1808260-84-5 FASN-IN-1 is a fatty acid synthase (FASN) inhibitor extracted from patent WO2015134790A1, compound 56 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2391 Supplier Medchemexpress LLC Supplier No. HY1117775MG
Medchemexpress, HY-111777 5mg FASN-IN-1 CAS:1808260-84-5 FASN-IN-1 is a fatty acid synthase (FASN) inhibitor extracted from patent WO2015134790A1, compound 56 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.