Learn More
Medchemexpress LLC HY-112829 5mg Medchemexpress, FASN-IN-2 CAS:1399177-37-7 Purity:>98%
SDP
Catalog No. 501932441
Encompass_Preferred
Medchemexpress, HY-112829 5mg FASN-IN-2 CAS:1399177-37-7 FASN-IN-2 is a Fatty Acid Synthase (FASN) inhibitor extracted from patent WO2012122391A1, compound 152, has an IC 50 of 0.052 μM and an EC 50 of 0.072 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2441 Supplier Medchemexpress LLC Supplier No. HY1128295MG
Medchemexpress, HY-112829 5mg FASN-IN-2 CAS:1399177-37-7 FASN-IN-2 is a Fatty Acid Synthase (FASN) inhibitor extracted from patent WO2012122391A1, compound 152, has an IC 50 of 0.052 μM and an EC 50 of 0.072 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.