Learn More
Medchemexpress LLC FASN-IN-3 | 2097262-60-5 | 384.47 | 10 MG
SDP
Catalog No. 5000448998
Encompass_Preferred
FASN-IN-3 is a fatty acid synthase (FASN) inhibitor. It was extracted from patent US20170119786A1, compound 242A. In vitro studies showed that FASN-IN-3 treatment of PBMC reduces IL-1 beta secretion from LPS or LTA stimulation. For monocytes, it reduces IL-1 beta from LTA stimulation, with a slight reduction after LPS stimulation.
  • Inhibits fatty acid synthase (FASN)
  • Reduces IL-1 beta secretion from PBMC stimulated with LPS or LTA
  • Reduces IL-1 beta from monocyte LTA stimulation
  • Solid appearance, off-white to yellow color
Catalog No. 50-004-48998 Supplier Medchemexpress LLC Supplier No. HYU0043610MG
FASN-IN-3 is a fatty acid synthase (FASN) inhibitor. It was extracted from patent US20170119786A1, compound 242A. In vitro studies showed that FASN-IN-3 treatment of PBMC reduces IL-1 beta secretion from LPS or LTA stimulation. For monocytes, it reduces IL-1 beta from LTA stimulation, with a slight reduction after LPS stimulation.
  • Inhibits fatty acid synthase (FASN)
  • Reduces IL-1 beta secretion from PBMC stimulated with LPS or LTA
  • Reduces IL-1 beta from monocyte LTA stimulation
  • Solid appearance, off-white to yellow color