Learn More
Abcam FASP Protein Digestion Kit, 8PACK
Catalog No. 501963756
Encompass_Preferred
FASP Protein Digestion Kit (ab270519) is for researchers who wish to solubilize whole or fractionated protein samples in SDS, digest the protein with trypsin, and analyse the resulting peptides by mass spectrometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-196-3756 Supplier Abcam Supplier No. AB2705198PACK
FASP Protein Digestion Kit (ab270519) is for researchers who wish to solubilize whole or fractionated protein samples in SDS, digest the protein with trypsin, and analyse the resulting peptides by mass spectrometry.
The product is subject to the following: Abcam Restricted Use Statement