Learn More
Medchemexpress LLC HY-P7168 10ug Medchemexpress, FASR/TNFRSF6 (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP716810UG
Encompass_Preferred
Medchemexpress, HY-P7168 10ug FASR/TNFRSF6 (HEK293-expressed), Human CAS: Recombinant Human FASR/TNFRSF6 (HEK293-expressed) belongs to the subgroup of the tumor necrosis factor receptor (TNF-R) family that can trigger apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3119