Learn More
Meridian Life Science Fast 1-Step RT-qPCR Reaction, 100ML
SDP
Catalog No. 5000856493
Encompass_Preferred
Fast 1-Step RT-qPCR Reaction Mix, 2x has been designed with the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with a fast hot-start polymerase, dNTPs and MgCl2, for highly reproducible, accurate, sensitive RT-qPCR efficiency in a single tube, even from low abundance targets and scarce samples. With the addition of reverse transcriptase and RNase Inhibitor, it delivers excellent results in both singleplex and multiplex RT-qPCR assays and confers superior assay performance under fast thermal cycling conditions.
Catalog No. 50-008-56493 Supplier Meridian Life Science Supplier No. MDX032100B
Fast 1-Step RT-qPCR Reaction Mix, 2x has been designed with the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with a fast hot-start polymerase, dNTPs and MgCl2, for highly reproducible, accurate, sensitive RT-qPCR efficiency in a single tube, even from low abundance targets and scarce samples. With the addition of reverse transcriptase and RNase Inhibitor, it delivers excellent results in both singleplex and multiplex RT-qPCR assays and confers superior assay performance under fast thermal cycling conditions.