Learn More
Perkin Elmer US LLC FAST 1 Valve Assembly
Catalog No. 502112178
Encompass_Preferred
This FAST 1 Valve is included in the NexION 2000 ICP-MS Productivity Configuration's Sample Introduction Kit (N8150036).
Catalog No. 50-211-2178 Supplier Perkin Elmer US LLC Supplier No. N8152359
This FAST 1 Valve is included in the NexION 2000 ICP-MS Productivity Configuration's Sample Introduction Kit (N8150036).