Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Blue BB Salt Hemi(Zinc Chloride) Salt, Sigma-Aldrich

Catalog No. 01258839 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-839 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-839 Supplier MilliporeSigma Supelco Supplier No. F337825G
Add to Cart
Add to Cart

Fast blue BB salt is suitable for the staining of enterochromaffin. Its applications are seen in enzyme histochemistry by Gomori and are suitable for the histochemical demonstration of beta-glucuronidase activities, and various peptidases and proteases. It is also used in enzyme histochemistry including the demonstration of serine esterase in blood smears and histological sections. Fast blue BB salt plays a vital role as a routine stain to demonstrate alkaline phosphatase labeled antibodies.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥80 %
Quantity 25 g
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.