Learn More
Fast Blue RR Salt, Sigma-Aldrich
Catalog No. 01258943 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
01-258-943 25 g
1 options
Catalog No. 01-258-943 Supplier MilliporeSigma Supelco Supplier No. F050025G
Add to Cart
Add to Cart
Histological staining reagent used to detect alkaline phosphatase and esterase activity.
Fast blue RR is mainly used in histological stainings. It binds to the reaction product of alkaline phosphatase and precipitates it as an insoluble compound. The sites of alkaline phosphatase activity appears black. Fast blue RR is also used for the detection of esterase activity. It couples with naphthol (released from naphthol acetate) and forms an insoluble azo-dye which appears red/blue.
TRUSTED_SUSTAINABILITY
Specifications
Form Crystalline
Quantity 25 g
Product Type Stain Kit